(RS)-Fmoc-α-methoxyglycine

Supplier: Matrix Scientific

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methoxyacetic acid

041405-250MG 041405-1G
101851-644EA 238.46 USD
101851-644 101851-646
(RS)-Fmoc-α-methoxyglycine
(RS)-Fmoc-alpha-methoxyglycine

Hazard: Irritant

Formula: C₁₈H₁₇NO₅
MW: 327.34 g/mol
MDL Number: MFCD05663764
CAS Number: 156059-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR