Fmoc-trans-4-aminocyclohexane acetic acid

Supplier: Matrix Scientific

041403-1G 041403-5G
101851-638EA 386.15 USD
101851-638 101851-640
Fmoc-trans-4-aminocyclohexane acetic acid
Fmoc-trans-4-aminocyclohexane acetic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MDL Number: MFCD04973171
CAS Number: 1217650-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR