Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-trans-4-Amc-OH, trans-4-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)cyclohexanecarboxylic acid, N-Fmoc-tranexamic acid, trans-4-[[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]methyl]cyclohexanecarboxylic acid, trans-4-(Fmoc-aminomethyl)cyclohexanecarboxylic acid

041402-1G 041402-5G
101851-634EA 224.62 USD
101851-634 101851-636
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MW: 379.46 g/mol
MDL Number: MFCD00273480
CAS Number: 167690-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR