Fmoc-L-2-amino-6-(O,O'-diethylphosphono)hexanoicacid
Supplier: Matrix Scientific
041401-100MG
041401-250MG
101851-630EA
686.15
USD
101851-630
101851-632
Fmoc-L-2-amino-6-(O,O'-diethylphosphono)hexanoicacid
Fmoc-L-2-amino-6-(O,O'-diethylphosphono)hexanoicacid
Hazard: Irritant
Formula:
C₂₅H₃₂NO₇P |
MDL Number:
MFCD09750535 CAS Number: 1217805-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...