Fmoc-DL-2-amino-1,3-dihydrophenalene-2-carboxylic acid
Supplier: Matrix Scientific
041400-250MG
041400-1G
101851-626EA
412.31
USD
101851-626
101851-628
Fmoc-DL-2-amino-1,3-dihydrophenalene-2-carboxylic acid
Fmoc-DL-2-amino-1,3-dihydrophenalene-2-carboxylic acid
Hazard: Irritant
Formula:
C₂₉H₂₃NO₄ |
MDL Number:
MFCD09750533 CAS Number: 1185296-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...