Fmoc-cis-4-aminocyclohexane acetic acid

Supplier: Matrix Scientific

041397-1G 041397-5G
101851-614EA 453.85 USD
101851-614 101851-616
Fmoc-cis-4-aminocyclohexane acetic acid
Fmoc-cis-4-aminocyclohexane acetic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MW: 379.46 g/mol
MDL Number: MFCD01862346
CAS Number: 1217675-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR