N-Fmoc-8-aminooctanoic acid

Supplier: Matrix Scientific

041396-1G 041396-5G
101851-610EA 123.08 USD
101851-610 101851-612
N-Fmoc-8-aminooctanoic acid
N-Fmoc-8-aminooctanoic acid

Hazard: Irritant

Formula: C₂₃H₂₇NO₄
MDL Number: MFCD00235890
CAS Number: 126631-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR