Fmoc-5-aminolevulinic acid

Supplier: Matrix Scientific

041393-1G 041393-5G
101851-598EA 630.77 USD
101851-598 101851-600
Fmoc-5-aminolevulinic acid
Fmoc-5-aminolevulinic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₅
MDL Number: MFCD02682636
CAS Number: 160111-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR