Fmoc-4-amino-tetrahydropyran-4-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid
041390-1G
041390-5G
101851-586EA
492.31
USD
101851-586
101851-588
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid
Hazard: Irritant
Formula:
C₂₁H₂₁NO₅ MW: 367.41 g/mol |
MDL Number:
MFCD02683137 CAS Number: 285996-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...