Fmoc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

041388-100MG 041388-500MG 041388-1G
101851-574EA 386.15 USD
101851-574 101851-576 101851-578
Fmoc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid
Fmoc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₁NO₄
MW: 375.42 g/mol
MDL Number: MFCD02682618
CAS Number: 1217848-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR