3-(endo-9-Fluorenylmethoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid

041386-100MG 041386-500MG 041386-1G
101851-564EA 386.15 USD
101851-564 101851-566 101851-568
3-(endo-9-Fluorenylmethoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid
3-(endo-9-Fluorenylmethoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₁NO₄
MW: 375.417 g/mol
MDL Number: MFCD02682616
CAS Number: 1212066-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR