Fmoc-2-aminooctanoic acid
Supplier: Matrix Scientific
041383-1G
041383-5G
101851-556EA
386.15
USD
101851-556
101851-558
Fmoc-2-aminooctanoic acid
Fmoc-2-aminooctanoic acid
Hazard: Irritant
Formula:
C₂₃H₂₇NO₄ |
MDL Number:
MFCD02094482 CAS Number: 197384-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...