Fmoc-2-aminoheptanoic acid

Supplier: Matrix Scientific

041382-1G 041382-5G
101851-552EA 598.46 USD
101851-552 101851-554
Fmoc-2-aminoheptanoic acid
Fmoc-2-aminoheptanoic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₄
MDL Number: MFCD02682634
CAS Number: 1219184-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR