Fmoc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)
Supplier: Matrix Scientific
041381-100MG
041381-1G
101851-548EA
275.38
USD
101851-548
101851-550
Fmoc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)
Fmoc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)
Hazard: Irritant
Formula:
C₂₃H₂₃NO₄ |
MDL Number:
MFCD12547706 CAS Number: 188751-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...