1-(Fmoc-amino)cyclopropanecarboxylic acid
Supplier: Matrix Scientific
Synonyms:
1-Aminocyclopropane-1-carboxylic acid, N-FMOC protected, Fmoc-ACPC-OH, Fmoc-1-aminocyclopropane-1-carboxylic acid, 1-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopropanecarboxylic acid
041379-1G
041379-5G
101851-540EA
107.69
USD
101851-540
101851-542
1-(Fmoc-amino)cyclopropanecarboxylic acid
1-(Fmoc-amino)cyclopropanecarboxylic acid
Hazard: Irritant
Formula:
C₁₉H₁₇NO₄ MW: 323.348 g/mol |
MDL Number:
MFCD00190873 CAS Number: 126705-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...