1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
Fmoc-cycloleucine, Fmoc-Cle-OH, 1-(Fmoc-amino)cyclopentanecarboxylic Acid
041378-1G
041378-5G
101851-536EA
44.62
USD
101851-536
101851-538
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid ≥95%
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic acid
Hazard: Irritant
Formula:
C₂₁H₂₁NO₄ MW: 351.40 g/mol |
MDL Number:
MFCD01074696 CAS Number: 117322-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...