Fmoc-1-aminocyclohexane-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(Fmoc-amino)cyclohexanecarboxylic Acid

041377-1G 041377-5G
101851-532EA 98.46 USD
101851-532 101851-534
Fmoc-1-aminocyclohexane-1-carboxylic acid
Fmoc-1-aminocyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₃NO₄
MW: 365.43 g/mol
MDL Number: MFCD00273464
CAS Number: 162648-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR