Fmoc-1-aminocyclobutane-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclobutanecarboxylic acid

041376-1G 041376-5G
101851-528EA 412.31 USD
101851-528 101851-530
Fmoc-1-aminocyclobutane-1-carboxylic acid
Fmoc-1-aminocyclobutane-1-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD02682624
CAS Number: 885951-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR