Fmoc-11-aminoundecanoic acid

Supplier: Matrix Scientific

041375-1G 041375-5G
101851-524EA 164.62 USD
101851-524 101851-526
Fmoc-11-aminoundecanoic acid
Fmoc-11-aminoundecanoic acid

Hazard: Irritant

Formula: C₂₆H₃₃NO₄
MDL Number: MFCD00235891
CAS Number: 88574-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR