trans-4-(Boc-amino)cyclohexaneacetic acid

Supplier: Matrix Scientific

Synonyms: 2-(trans-4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid, Boc-1,4-trans-acha-oh

041367-1G 041367-5G
101851-500EA 275.38 USD
101851-500 101851-502
trans-4-(Boc-amino)cyclohexaneacetic acid
trans-4-(Boc-amino)cyclohexaneacetic acid

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD03844588
CAS Number: 189153-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR