trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid
Supplier: Matrix Scientific
Synonyms:
trans-4-[(Boc-amino)methyl]cyclohexanecarboxylic Acid, trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid
041366-1G
041366-5G
101851-496EA
15.38
USD
101851-496
101851-498
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid
Hazard: Irritant/Store in Freezer
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol |
MDL Number:
MFCD00673773 CAS Number: 27687-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...