Boc-L-2-amino-6-(O,O'-diethylphosphono)hexanoic acid

Supplier: Matrix Scientific

041365-100MG 041365-250MG
101851-492EA 686.15 USD
101851-492 101851-494
Boc-L-2-amino-6-(O,O'-diethylphosphono)hexanoic acid
Boc-L-2-amino-6-(O,O'-diethylphosphono)hexanoic acid

Hazard: Irritant

Formula: C₁₅H₃₀NO₇P
MDL Number: MFCD09750534
CAS Number: 1217810-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR