Boc-DL-2-amino-1,3-dihydrophenalene-2-carboxylicacid
Supplier: Matrix Scientific
041364-250MG
041364-1G
101851-488EA
412.31
USD
101851-488
101851-490
Boc-DL-2-amino-1,3-dihydrophenalene-2-carboxylicacid
Boc-DL-2-amino-1,3-dihydrophenalene-2-carboxylicacid
Hazard: Irritant
Formula:
C₁₉H₂₁NO₄ |
MDL Number:
MFCD09750532 CAS Number: 1185302-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...