cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Supplier: Matrix Scientific

Synonyms: cis-4-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, cis-4-(BOC-amino)cyclohexanecarboxylic acid, cis-4-(Boc-amino)cyclohexanecarboxylic Acid

041362-1G 041362-5G
101851-484EA 92.31 USD
101851-484 101851-486
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
MDL Number: MFCD01862294
CAS Number: 53292-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR