cis-4-(Boc-amino)cyclohexaneacetic acid
Supplier: Matrix Scientific
Synonyms:
cis-[4-(Boc-amino)cyclohexyl]acetic acid, 2-(cis-4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid
041361-1G
041361-5G
101851-480EA
246.15
USD
101851-480
101851-482
cis-4-(Boc-amino)cyclohexaneacetic acid
cis-4-(Boc-amino)cyclohexaneacetic acid
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ |
MDL Number:
MFCD01862296 CAS Number: 327156-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...