Boc-8-aminooctanoic acid
Supplier: Matrix Scientific
Synonyms:
Boc-8-Aoc-OH
041360-1G
041360-5G
101851-476EA
60
USD
101851-476
101851-478
Boc-8-aminooctanoic acid
Boc-8-aminooctanoic acid
Hazard: Irritant
Formula:
C₁₃H₂₅NO₄ MW: 259.35 g/mol |
MDL Number:
MFCD00270350 CAS Number: 30100-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...