N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid

Supplier: Matrix Scientific

041359-1G 041359-5G
101851-472EA 36.92 USD
101851-472 101851-474
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD00063356
CAS Number: 60142-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR