N-(tert-Butoxycarbonyl)-5-aminovaleric acid

Supplier: Matrix Scientific

Synonyms: Boc-5-Ava-OH, BOC-5-Aminopentanoic acid, 5-(Boc-amino)valeric Acid, N-(tert-Butoxycarbonyl)-5-aminopentanoic Acid, 5-(Boc-amino)pentanoic Acid, N-Boc-5-aminovaleric Acid, 5-[(tert-Butoxycarbonyl)amino]valeric Acid, Boc-NH-C4-acid, N-Boc-5-aminopentanoic Acid, 5-[(tert-Butoxycarbonyl)amino]pentanoic Acid

041358-1G 041358-5G
101851-468EA 84.62 USD
101851-468 101851-470
N-(tert-Butoxycarbonyl)-5-aminovaleric acid
N-(tert-Butoxycarbonyl)-5-aminovaleric acid

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
MDL Number: MFCD00076903
CAS Number: 27219-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR