Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid

Supplier: Matrix Scientific

Synonyms: diexo-3-tert-Butoxycarbonylamino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

041354-100MG 041354-500MG 041354-1G
101851-448EA 386.15 USD
101851-448 101851-450 101851-452
Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid
Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄
MW: 253.3 g/mol
MDL Number: MFCD02682617
CAS Number: 148257-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR