3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected

Supplier: Matrix Scientific

Synonyms: Boc-3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid

041353-100MG 041353-500MG
101851-444EA 386.15 USD
101851-444 101851-446
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.31 g/mol
MDL Number: MFCD02682613
CAS Number: 1212373-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR