Boc-3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid

Supplier: Matrix Scientific

041352-100MG 041352-500MG 041352-1G
101851-438EA 350.77 USD
101851-438 101851-440 101851-442
Boc-3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid
Boc-3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄
MW: 253.3 g/mol
MDL Number: MFCD02682615
CAS Number: 148257-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR