[(tert-Butoxycarbonyl)aminooxy]acetic acid

Supplier: Matrix Scientific

Synonyms: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

041351-1G 041351-5G
101851-434EA 46.15 USD
101851-434 101851-436
[(tert-Butoxycarbonyl)aminooxy]acetic acid
[(tert-Butoxycarbonyl)aminooxy]acetic acid

Hazard: Irritant

Formula: C₇H₁₃NO₅
MW: 191.18 g/mol
MDL Number: MFCD01632027
CAS Number: 42989-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR