Boc-2-aminooctanoic acid

Supplier: Matrix Scientific

041349-1G 041349-5G
101851-430EA 538.46 USD
101851-430 101851-432
Boc-2-aminooctanoic acid
Boc-2-aminooctanoic acid

Hazard: Irritant

Formula: C₁₃H₂₅NO₄
MDL Number: MFCD02094483
CAS Number: 14676-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR