Boc-2-aminoheptanoic acid

Supplier: Matrix Scientific

041348-1G 041348-5G
101851-426EA 598.46 USD
101851-426 101851-428
Boc-2-aminoheptanoic acid
Boc-2-aminoheptanoic acid

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MDL Number: MFCD00270724
CAS Number: 64769-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR