Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)bicyclo[2.2.1]heptane-2-carboxylic acid

041347-100MG 041347-1G
101851-422EA 275.38 USD
101851-422 101851-424
Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)
Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD25132076
CAS Number: 172927-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR