Boc-2-amino-2-indancarboxylic acid

Supplier: Matrix Scientific

041346-250MG 041346-1G
101851-418EA 504.62 USD
101851-418 101851-420
Boc-2-amino-2-indancarboxylic acid
Boc-2-amino-2-indancarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
MDL Number: MFCD00800597
CAS Number: 71066-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR