Boc-cycloleucine

Supplier: Matrix Scientific

Synonyms: 1-(Boc-amino)cyclopentanecarboxylic acid, 1-N-BOC-Aminocyclopentanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

041344-1G 041344-5G
101851-410EA 20 USD
101851-410 101851-412
Boc-cycloleucine
Boc-cycloleucine

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD01076126
CAS Number: 35264-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR