Boc-1-aminocyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, 1-[(tert-Butoxycarbonyl)amino]cyclohexanecarboxylic acid, 1-(Boc-amino)cyclohexanecarboxylic Acid

041343-1G 041343-5G
101851-406EA 21.54 USD
101851-406 101851-408
Boc-1-aminocyclohexane-1-carboxylic acid ≥95%
Boc-1-aminocyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD00273427
CAS Number: 115951-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR