1-(Boc-amino)cyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

041342-1G 041342-5G
101851-402EA 86.15 USD
101851-402 101851-404
1-(Boc-amino)cyclobutanecarboxylic acid ≥95%
1-(Boc-amino)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
MDL Number: MFCD02682623
CAS Number: 120728-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR