Boc-11-Aun-OH

Supplier: Matrix Scientific

Synonyms: 11-[(tert-Butoxycarbonyl)amino]undecanoic acid, 11-(Boc-amino)-undecanoic acid, 11-(Boc-amino)undecanoic acid, Boc-11-Aun-OH, Boc-11-aminoundecanoic acid

041341-1G 041341-5G
101851-398EA 76.92 USD
101851-398 101851-400
Boc-11-Aun-OH
Boc-11-Aun-OH

Hazard: Irritant

Formula: C₁₆H₃₁NO₄
MW: 301.427 g/mol
MDL Number: MFCD00076902
CAS Number: 10436-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR