Bis(boc-aminooxy)acetic acid

Supplier: Matrix Scientific

Synonyms: ((Bis((1,1-dimethylethoxy)carbonyl)amino)oxy)acetic acid

041334-1G 041334-5G
101851-378EA 363.08 USD
101851-378 101851-380
Bis(boc-aminooxy)acetic acid
Bis(boc-aminooxy)acetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₇
MDL Number: MFCD06410966
CAS Number: 293302-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR