(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(1S,3R)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-FMOC protected, (1S,3R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopentanecarboxylic acid
041331-250MG
041331-1G
101851-366EA
147.69
USD
101851-366
101851-368
(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid
(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid
Hazard: Irritant
Formula:
C₂₁H₂₁NO₄ MW: 351.41 g/mol |
MDL Number:
MFCD01311757 CAS Number: 220497-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...