(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, Boc-D-AcPAC, (1S,3R)-3-(Boc-amino)cyclopentanecarboxylic Acid, (1S,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
041330-250MG
041330-1G
101851-362EA
135.38
USD
101851-362
101851-364
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid ≥95%
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Hazard: Irritant/Refrigerate
Formula:
C₁₁H₁₉NO₄ |
MDL Number:
MFCD01320857 CAS Number: 261165-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...