(1S,2S)-Boc-Achc
Supplier: Matrix Scientific
Synonyms:
(1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid
041327-250MG
041327-1G
101851-350EA
312.31
USD
101851-350
101851-352
(1S,2S)-Boc-Achc
(1S,2S)-Boc-Achc
Hazard: Irritant
Formula:
C₁₂H₂₁NO₄ MW: 243.31 g/mol |
MDL Number:
MFCD04974226 CAS Number: 488703-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...