(1S,2S)-Boc-Acpc

Supplier: Matrix Scientific

Synonyms: (1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

041326-250MG 041326-1G
101851-346EA 90.77 USD
101851-346 101851-348
(1S,2S)-Boc-Acpc
(1S,2S)-Boc-Acpc

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD04974224
CAS Number: 143679-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR