(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid

Supplier: Matrix Scientific

Synonyms: (1R,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopent-2-enecarboxylic acid

041321-250MG 041321-1G
101851-334EA 300 USD
101851-334 101851-336
(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid
(1R,4S)-Fmoc-4-aminocyclopent-2-enecarboxylic acid

Hazard: Irritant

Formula: C₂₁H₁₉NO₄
MW: 349.39 g/mol
MDL Number: MFCD01311756
CAS Number: 220497-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR