(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (1R,4S)-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid

041320-250MG 041320-1G
101851-330EA 252.31 USD
101851-330 101851-332
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD00211288
CAS Number: 151907-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR