(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
041318-1G
041318-5G
101851-324EA
526.15
USD
101851-324
102663-702
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₉NO₄ |
MDL Number:
MFCD01320858 CAS Number: 161660-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...