(1R,2S)-Boc-Achc

Supplier: Matrix Scientific

Synonyms: (1R,2S)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

041312-250MG 041312-1G
101851-306EA 1855.38 USD
101851-306 101851-308
(1R,2S)-Boc-Achc
(1R,2S)-Boc-Achc

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD06410968
CAS Number: 352356-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR