(1R,2R)-Fmoc-Acpc

Supplier: Matrix Scientific

Synonyms: (1R,2R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopentanecarboxylic acid

041307-250MG 041307-1G
101851-294EA 263.08 USD
101851-294 101851-296
(1R,2R)-Fmoc-Acpc
(1R,2R)-Fmoc-Acpc

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD04112693
CAS Number: 359586-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR